128-3000-00 Rev D - Thermo Scientific RVT100_RVT400_RVT4104 Refrigerated Vapor Trap - Instruction Manual
The Instruction Manual provides detailed guidance on the use, installation, and maintenance of the RVT-series traps, designed for solvent vapor condensation and collection. The manual covers model descriptions, site preparation, power requirements, ventilation needs, operation details, and safety protocols. These traps protect vacuum pumps from solvent vapors, differing in capacity and operating temperatures, with optional accessories for enhanced functionality.
Description:
Instruction manual
Environment:
Thermo Scientific® RVT100, RVT400, RVT4104 Refrigerated Vapor Trap:
| Cold Traps | RVT100 | RVT400 | RVT4104 |
| Operating Temperature* | -50 °C | -50 °C | -104 °C |
| Capacity | 1 Liter | 4 Liter | 4 Liter |
| Power Requirements | 120 VAC/60 Hz, 4 A | 120 VAC/60 Hz, 4 A | 120 VAC/60 Hz, 12 A |
| 240 VAC/50 Hz, 2 A | 240 VAC/50 Hz, 2 A | 240 VAC/50 Hz, 6 A | |
| * Depends on ambient temperature, line voltage fluctuations, and load capacity. | |||
RVT100
RVT400
RVT4104
Attachment(s):
| File | Last Modified |
|---|---|
| 128-3000-00 - Rev D - Thermo Scientific RVT100, RVT400, RVT4104 Refrigerated Vapor Trap - Instruction Manual.pdf | January 03, 2022 |