128-3000-51 - RVT5105 Refrigerated Vapor Traps - Installation and Operation Manual
The Operation Manual provides detailed instructions on the installation, operation, and maintenance of the RVT5105. These traps are designed for solvent vapor condensation and collection, protecting high-vacuum rotary vane oil pumps. The manual covers receiving and inspection, installation requirements, operational guidelines, and troubleshooting tips. It also highlights necessary accessories such as Glass Condensation Flasks and CryoCool Heat Transfer Fluid.
Description:
Installation and operation manual
Environment:
Thermo Scientific RVT5105 Refrigerated Vapor Traps, power requirements:
- 115 VAC/60 Hz, 13A
- 230 VAC/50 Hz, 7A
- RVT5105-115
- RVT5105-220
- RVT 5105
- RVT-5105
Attachment(s):
| File | Last Modified |
|---|---|
| 128-3000-51 - RVT5105 Refrigerated Vapor Traps - Installation and Operation Manual.pdf | January 03, 2022 |