133-3001-00 Rev D - Thermo Scientific SPD1010 and SPD2010 Integrated SpeedVac System - Instruction Manual
The Thermo Scientific SPD1010 and SPD2010 Integrated SpeedVac Systems instruction manual provides detailed guidance on the operation, installation, and troubleshooting of these solvent evaporation, sample concentration, and drying systems. The SPD1010 and SPD2010 combine centrifugal force, vacuum, and applied heat to prevent sample bumping and foaming. The SPD2010 processes up to four 500 ml samples, while the SPD1010 handles up to four 100 ml samples.
Description:
Instruction manual
Environment:
Thermo Scientific SPD1010/SPD2010 Integrated SpeedVac® System:
| Model: | SPD1010 | SPD2010 |
| Operative Power: | 115 VAC; 60 Hz;12 amps | 220 VAC; 60 Hz; 8 amps |
| 230 VAC; 50 Hz; 6 amps | 230 VAC; 50 Hz; 8 amps |
SPD1010
SPD-1010
SPD 1010
SPD2010
SPD-2010
SPD 2010
*Dependent upon ambient temperature, line voltage fluctuation, and load capacity.
Attachment(s):
| File | Last Modified |
|---|---|
| 133-3001-00 - Rev D - Thermo Scientific SPD1010 and SPD2010 Integrated SpeedVac System - Instruction Manual.pdf | January 03, 2022 |