145-3001-00 Rev C - Universal Vacuum System UVS400 and UVS400A - Instruction Manual
The attached content describes the Thermo Electron Universal Vacuum System (UVS400/UVS400A), a versatile vacuum source and solvent recovery system. Its key components include: a refrigerated vapor trap, oil-free vacuum pump, and VaporNet controller. The instrument is characterized by its low maintenance, reliability, energy efficiency, and environmental safety. It is suitable for various laboratory applications and able to recover evaporated solvents entirely.
Description:
UVS400 and UVS400A User Manual
Environment:
- UVS400 (also known as UVS 400, UVS-400)
- UVS400A (also known as UVS 400A, UVS-400A)
Attachment(s):
| File | Last Modified |
|---|---|
| UVS400.PDF | April 16, 2024 |