197-3003-00 Rev G - Savant SPD131DDA SpeedVac Concentrator - Instruction Manual
The instruction manual for the Thermo Scientific Savant SPD131DDA SpeedVac Concentrator provides detailed guidance on the installation, operation, and maintenance of the unit. Key sections include an overview of the SPD131DDA unit, control panel descriptions, manual and auto run functionalities, rotor selection guidelines, solvent choice recommendations, and maintenance procedures. Additionally, it covers specifications and RS232 port specifications for comprehensive user support.
Description:
Instruction manual
Environment:
Thermo Scientific Savant® SpeedVac Concentrator, model SPD131DDA. Power requirements:
- 115 VAC/60Hz, 5.0A (Part number SPD131 DDA-115)
- 230 VAC/50Hz, 3.0A (Part number SPD131 DDA-230)
- SPD131 DDA
Attachment(s):
| File | Last Modified |
|---|---|
| 197-3003-00 - Rev G - Savant SPD131DDA SpeedVac Concentrator - Instruction Manual.pdf | January 03, 2022 |