Thermo Fisher Scientific
80302051 - Rev C - SpeedVac SPD1030 and SPD2030 Vacuum Concentrator - Installation and Operation Manual
Description: 
Installation and operation manual
Environment:
SpeedVac™ SPD1030 and SPD2030 Vacuum Concentrator:
  SPD1030  SPD2030
Capacity Moderate Large
Operative Power* 115 VAC; 60 Hz; 12 amps 220 VAC; 60 Hz; 8 amps
  230 VAC; 50 Hz; 6 amps 230 VAC; 50 Hz; 8 amps
SPD1030
SPD 1030
SPD2030
SPD 2030
*Dependent upon ambient temperature, line voltage fluctuation, and load capacity.