80302051 Rev C - SPD1030 and SPD2030 Vacuum Concentrator - Instruction Manual
This manual provides comprehensive guidance on the installation and operation of SpeedVac SPD1030 and SPD2030 Vacuum Concentrators. It covers safety precautions, operating standards, product specifications, and environmental conditions necessary for optimal performance. The manual emphasizes the importance of qualified personnel for internal adjustments and maintenance to ensure safety and functionality.
Description:
Installation and operation manual
Environment:
SpeedVac™ SPD1030 and SPD2030 Vacuum Concentrator:
| SPD1030 | SPD2030 | |
| Capacity | Moderate | Large |
| Operative Power* | 115 VAC; 60 Hz; 12 amps | 220 VAC; 60 Hz; 8 amps |
| 230 VAC; 50 Hz; 6 amps | 230 VAC; 50 Hz; 8 amps |
SPD1030
SPD 1030
SPD2030
SPD 2030
*Dependent upon ambient temperature, line voltage fluctuation, and load capacity.
Attachment(s):
| File | Last Modified |
|---|---|
| 80302051 - Rev C - SpeedVac SPD1030 and 2030 Vacuum Concentrator - Installation and Operation Manual.pdf | January 03, 2022 |