80302105 Rev A - RVT450 Refrigerated Vapor Trap - Installation and Operation Manual
This manual provides comprehensive guidelines on the setup, usage, and maintenance of the RVT450 Refrigerated Vapor Trap. It covers the product's purpose to condense and collect solvent vapors, replacing dry ice/methanol traps, and protecting vacuum pumps. Key sections include safety precautions, product specifications, installation steps, operational instructions, and warranty details, ensuring safe and efficient operation of the device.
Description:
Installation and operation manual
Environment:
RVT450 Refrigerated Vapor Trap, operative power*:
- 115 VAC; 60 Hz; 8 amps
- 230 VAC; 50 Hz; 6 amps
- Savant
- RVT450
- RVT 450
*Dependent upon ambient temperature, line voltage fluctuation, and load capacity.
Attachment(s):
| File | Last Modified |
|---|---|
| 80302105 - Rev A - RVT450 Refrigerated Vapor Trap - Installation and Operation Manual.pdf | January 03, 2022 |