80302106 Rev A - UVS850DDA Universal Vacuum System with Vapornet - Installation and Operation Manual
This manual provides comprehensive guidelines for installing and operating the UVS850DDA Universal Vacuum System. It includes safety precautions, operating standards, system specifications, and detailed installation procedures. This system features a Vapornet Controller, TEFLON® tubing, and a high-efficiency oil-free pump for effective solvent recovery and vapor management. The manual ensures proper setup and maintenance for optimal performance.
Description:
Installation and operation manual
Environment:
UVS850DDA Universal Vacuum System with Vapornet, operative power*:
- 115 VAC; 60 Hz; 8 amps
- 230 VAC; 50 Hz; 6 amps
*Dependent upon ambient temperature, line voltage fluctuation, and load capacity.
Attachment(s):
| File | Last Modified |
|---|---|
| 80302106 - Rev A - UVS850DDA Universal Vacuum System with Vapornet - Installation and Operation Manual.pdf | January 03, 2022 |