80302107 Rev A - UVS450 Universal Vacuum System - Installation and Operation Manual
This manual provides detailed guidance on the setup, operation, and maintenance of the UVS450 system. It covers safety precautions, product specifications, installation steps, and operational instructions, including the use of the Vapornet feature for solvent recovery. Maintenance tips, such as checking and emptying the Glass Condensation Flask, are also included to ensure efficient performance.
Description:
Installation and operation manual
Environment:
Thermo Scientific Universal Vacuum System:
| Model | Operative Power* |
| UVS450 | 115 VAC; 60 Hz; 8 amps |
| UVS450A | 230 VAC; 50 Hz; 6 amps |
*Dependent upon ambient temperature, line voltage fluctuation, and load capacity.
Attachment(s):
| File | Last Modified |
|---|---|
| 80302107 - Rev A - UVS450 Universal Vacuum System - Installation and Operation Manual.pdf | January 03, 2022 |