80302120 Rev A - SpeedVac SPD300DDA Vacuum Concentrator - Installation and Operation Manual
This manual provides comprehensive guidance on setting up and using the Thermo Scientific Savant SpeedVac SPD300DDA vacuum concentrator. It covers safety precautions, operating standards, product specifications, and detailed installation instructions. The manual also includes steps for operation, maintenance, and recommended accessories for optimal performance. This ensures efficient batch processing of samples with various solvents.
Description:
Installation and operation manual
Environment:
SpeedVac™ SPD300DDA Vacuum Concentrator, operative power*:
- 115 VAC; 60 Hz; 10 amps
- 230 VAC; 50 Hz; 6 amps
*Dependent upon ambient temperature, line voltage fluctuation, and load capacity.
Attachment(s):
| File | Last Modified |
|---|---|
| 80302120 - Rev A - SpeedVac SPD300DDA Vacuum Concentrator - Installation and Operation Manual.pdf | January 03, 2022 |