80302131 Rev A - SpeedVac SPD210 Vacuum Concentrator - Installation and Operation
The SpeedVac SPD210 Vacuum Concentrator is designed for efficient sample processing by removing non-aggressive organics such as methanol, acetonitrile, and water. Installation requires placing the unit with proper clearance, ensuring electrical compatibility, and setting up a vapor trap between the concentrator and vacuum pump. Operationally, it features microprocessor control for automatic or manual run conditions, with safety precautions requiring qualified personnel for maintenance.
Description:
Installation and operation
Environment:
Thermo Scientific Savant SpeedVac™ SPD210 vacuum concentrator
SPD210
SPD 210
Attachment(s):
| File | Last Modified |
|---|---|
| 80302131 - Rev A - SpeedVac SPD210 Vacuum Concentrator - Installation and Operation.pdf | December 29, 2021 |