Thermo Fisher Scientific
7018877 - 18" Forma Fury Washer - Instruction Manual
Description: 
Instruction manual
Environment:
18" Forma Fury Washer, model numbers and electrical requirements:
Model Electrical Requirements
8877 208/230 VAC, 1 Phase, 3 Wire (Requires Neutral) (Steam), 15 FLA Nominal
8878 208/230 VAC, 1 Phase, 3 Wire (Requires Neutral) (Electric), 60 Hz, 54 FLA Nominal
8879 208/230 VAC, 3 Phase, 4 Wire (Requires Neutral) (Electric), 60 Hz. 34 FLA Nominal
8803 220/240 VAC, 1 Phase, 2 Wire (Steam), 50 Hz
8808 380 VAC, 3 Phase, 4 Wire (Requires Neutral) (Electric), 50 Hz
NOTE: Full Load Amps (FLA) will vary, depending upon the line voltage and motor loading.