7018877 - 18" Forma Fury Washer - Instruction Manual
The attached document is the Instruction Manual for the 18" Forma Fury Washer (models 8877, 8878, 8879, 8805, and 8808) and contains these sections: Introduction, Specifications, Operation, installation & Start-Up, Routine Maintenance, and Service. Supplemental information includes parts lists, schematics, and warranty information. The attachment is a scanned copy of the original manual. Text cannot be copied from the manual.
Description:
Instruction manual
Environment:
18" Forma Fury Washer, model numbers and electrical requirements:
| Model | Electrical Requirements |
| 8877 | 208/230 VAC, 1 Phase, 3 Wire (Requires Neutral) (Steam), 15 FLA Nominal |
| 8878 | 208/230 VAC, 1 Phase, 3 Wire (Requires Neutral) (Electric), 60 Hz, 54 FLA Nominal |
| 8879 | 208/230 VAC, 3 Phase, 4 Wire (Requires Neutral) (Electric), 60 Hz. 34 FLA Nominal |
| 8803 | 220/240 VAC, 1 Phase, 2 Wire (Steam), 50 Hz |
| 8808 | 380 VAC, 3 Phase, 4 Wire (Requires Neutral) (Electric), 50 Hz |
NOTE: Full Load Amps (FLA) will vary, depending upon the line voltage and motor loading.
Attachment(s):
| File | Last Modified |
|---|---|
| 7018877 - 18_ Forma Fury Washer - Instruction Manual.pdf | January 04, 2022 |