257312-001 0708 - Rev A - Optimizing HP Ammonia Readings
This article offers tips for optimizing ammonia measurements with the Thermo Scientific Orion High Performance Ammonia ISE (Cat. No. 9512HPBNWP or 9512HP01). It notes the importance of using specific filling solutions, membranes, and ISAs. Key practices include proper soaking, avoiding air bubbles, correct membrane placement, and proper calibration. Following these guidelines ensures accurate ammonia measurements.
Description:
Tips for using the High Performance Ammonia ion selective electrode optimally.
Environment:
- 9512HPBNWP (High-Performance Ammonia Electrode)
Attachment(s):
| File | Last Modified |
|---|---|
| 257312-001 0708 - Rev A - Optimizing HP Ammonia Readings.pdf | April 07, 2023 |