AN-CHLORINEXP-E 0714 - Rev A - AN 003 On-Line Chlorine Monitoring for Drinking Water Using US EPA Method 334.0
This application note evaluates the Thermo Scientific Orion Chlorine XP analyzer's compliance with US EPA Method 334.0 for on-line chlorine monitoring in drinking water. Comparisons with grab samples using the Thermo Scientific Orion AQ3070 AQUAfast Chlorine Meter showed Chlorine XP readings within 0.1 mg/L of grab samples, meeting EPA criteria. The Chlorine XP measures free and total chlorine, pH, and temperature, ensuring comprehensive water quality monitoring.
Description:
Application note for measuring On-Line Chlorine Monitoring for Drinking Water Using US EPA Method 334.0
Environment:
- Thermo Scientific Orion Chlorine XP Water Quality Analyzer (Orion CXP72PH)
- Thermo Scientific Orion AQ3070 AQUAfast Chlorine Meter (Orion AQ3070)
Attachment(s):
| File | Last Modified |
|---|---|
| AN-CHLORINEXP-E 0714 - Rev A - AN 003 On-Line Chlorine Monitoring for Drinking Water Using US EPA Method 334.0.pdf | May 22, 2024 |