AN-NWINE-E 0315 - Rev A - AN 019 Nitrogen Measurement in Wine
This application note from Thermo Fisher Scientific emphasizes the importance of nitrogen testing in winemaking. Nitrogen is crucial for yeast growth and fermentation, affecting wine quality. It details nitrogen compounds like Yeast Assimilable Nitrogen (YAN) and outlines methods for testing ammonia and amino nitrogen. Equipment recommendations for different testing needs are provided, underscoring the need for accurate nitrogen measurement to ensure successful fermentation and high-quality wine
Description:
Application note for Nitrogen Measurement in Wine
Environment:
Thermo Scientific Orion StarA214
Thermo Scientific Orion StarA324
Thermo Scientific Orion STARA211
Thermo Scientific Orion STARA221
Thermo Scientific Orion 9512HPBNWP High-Performance Ammonia Electrode
Thermo Scientific Orion ROSS 8172BNWP Sure-Flow pH electrode
Thermo Scientific Orion GD9156BNWP Green pH Combination electrode
Attachment(s):
| File | Last Modified |
|---|---|
| AN-NWINE-E 0315 - Rev A - AN 019 Nitrogen Measurement in Wine.pdf | October 29, 2024 |