COL25606 0422 - AN 013 Measuring Clarity in Wine
This article details the process of measuring wine turbidity using Thermo Scientific Orion AQUAfast turbidity meters. It covers the equipment and reagents needed, calibration verification, and steps for accurate turbidity measurements of red, white, and rosé wines. The Orion AQUAfast AQ3010 and AQ4500 meters provide reliable readings unaffected by wine color, with the AQ4500 also suitable for water turbidity testing. Results demonstrate the meters' effectiveness in winemaking stages
Description:
Application note for measuring clarity in wine.
Attachment(s):
| File | Last Modified |
|---|---|
| COL25606 0422 - AN 013 Measuring Clarity in Wine.pdf | January 23, 2023 |