COL26156 0622 - AN 020 Measuring pH in Soil
This application note details the process of measuring soil pH using the Orion Combination Sure-Flow pH Electrode, emphasizing the importance of soil pH for plant and crop suitability. The document covers equipment setup, including the Thermo Scientific Orion Star A211 Benchtop pH Meter and necessary electrodes, and outlines steps for calibration, sample preparation, and analysis. Quality control procedures and statistical results are also provided to ensure accurate and reliable measurements.
Description:
Application note for measuring pH in soil.
Environment:
- 9165BNWP (Sure-Flow® combination pH electrode with epoxy body)
Attachment(s):
| File | Last Modified |
|---|---|
| COL26156 0622 - AN 020 Measuring pH in Soil.pdf | December 28, 2022 |