Log 121 - Rev A - Tip ORP - mV RmV and Eh
This article from Thermo Fisher Scientific's Water Analysis Instruments division explains different units for measuring Oxidation-Reduction Potential (ORP) in water analysis. It covers ORP in millivolts (mV), converting ORP to Eh (standard hydrogen electrode potential), and using RmV (relative millivolts) for calibrated readings. The goal is to help users understand and accurately measure ORP in various applications
Description:
Application log for interpreting Oxidative Reductive Potential (ORP) in units of measure
Environment:
- Thermo Scientific Orion Epoxy Sure-Flow combination redox/ORP electrode (Orion 9678BNWP)
- Thermo Scientific Orion Glass combination redox/ORP electrode (Orion 9778BNWP)
- Thermo Scientific Orion Epoxy Refillable combination redox/ORP/ATC electrode (Orion 9180BNMD)
- Thermo Scientific Orion Epoxy Low Maintenance combination redox/ORP/ATC electrode (Orion 9179BNMD)
Attachment(s):
| File | Last Modified |
|---|---|
| Log 121 - Rev A - Tip ORP - mV, RmV, and Eh.pdf | December 27, 2024 |