Thermo Fisher Scientific
Log 124 - Rev A - Tip ORP Measurement Frequently Asked Questions
This article from Thermo Fisher Scientific's Water Analysis Instruments division covers FAQs on ORP measurement, focusing on standards, result interpretation, and probe maintenance. It recommends the Orion 967901 or 967961 standard for its stability and accuracy. Factors affecting ORP readings include filling solutions, temperature, response time, rinsing, and electrode surface condition. Recommended electrodes include the Orion 9678BNWP and 9778BNWP for various applications.
Description: 
Application log for frequently asked ORP questions
Environment:
  • Thermo Scientific Orion Epoxy Sure-Flow combination redox/ORP electrode (Orion 9678BNWP)
  • Thermo Scientific Orion Glass combination redox/ORP electrode (Orion 9778BNWP)
  • Thermo Scientific Orion Epoxy Refillable combination redox/ORP/ATC electrode (Orion 9180BNMD)
  • Thermo Scientific Orion Epoxy Low Maintenance combination redox/ORP/ATC electrode (Orion 9179BNMD)
  • Was this article helpful?