Log 30 - Rev A - Redox Basics - Ready for Redox
The article discusses the redox process in "pickling" steel using a solution of ferric sulfate (Fe(+3)) and sulfuric acid. Iron dissolves as ferrous sulfate (Fe(+2)) and is monitored with an ORP electrode, which measures the electron exchange between Fe(+2) and Fe(+3) to maintain a high ratio, preventing steel pitting. It explains the electrode's function and mentions other redox pairs with high exchange rates used in analytical chemistry, like iodide/iodine and silver/silver ion.
Description:
Application note for how redox and ORP works
Environment:
- Thermo Scientific Orion Epoxy Sure-Flow combination redox/ORP electrode (Orion 9678BNWP)
- Thermo Scientific Orion Glass combination redox/ORP electrode (Orion 9778BNWP)
- Thermo Scientific Orion Epoxy Refillable combination redox/ORP/ATC electrode (Orion 9180BNMD)
- Thermo Scientific Orion Epoxy Low Maintenance combination redox/ORP/ATC electrode (Orion 9179BNMD)
Attachment(s):
| File | Last Modified |
|---|---|
| Log 30 - Rev A - Redox Basics - Ready for Redox.pdf | December 27, 2024 |