TN-AMWATER-E 0615 - Rev B - Measuring Ammonia in Water and Wastewater using the Dual Star pH ISE Meter
This technical note from Thermo Fisher Scientific explains how to measure ammonia levels in water and wastewater using the Thermo Scientific Orion Dual Star pH/ISE meter. It outlines the environmental impact of ammonia, necessary equipment, electrode setup, calibration standard preparation, and sample measurement procedures. The guide aims to ensure accurate ammonia readings, crucial for wastewater management and environmental protection.
Description:
Technical note for using the Dual Star pH/ISE meter with an ammonia ion selective electrode for measuring water and wastewater.
Environment:
- 2115000 (Orion Dual Star pH/ISE Dual Channel Meter)
- 9512HPBNWP (High-Performance Ammonia Electrode)
Attachment(s):
| File | Last Modified |
|---|---|
| TN-AMWATER-E 0615 - Rev B - Measuring Ammonia in Water and Wastewater using the Dual Star pH ISE Meter.pdf | April 07, 2023 |