Thermo Fisher Scientific
TN Low Level Total Alkalinity per SM2320
This technical note outlines the procedure for measuring low-level total alkalinity in water using a Thermo Scientific Orion Star T9102 pH titrator. The method involves titrating with 0.02N sulfuric acid to pH endpoints of 4.5 and 4.2, then calculating alkalinity as mg/L CaCO3. It includes equipment and sample preparation instructions, programmed parameters for titration, and guidelines for standardizing the titrant for accuracy.
Description: 
Technical note for performing low level alkalinity with Thermo Scientific Orion Star T9102 according to Standard Method SM2320.
Environment:
START9102 Thermo Scientific Orion Star T910 pH Titrator kit
START9100 Thermo Scientific Orion Star T910 pH Titrator
  • Was this article helpful?