Thermo Fisher Scientific
STARA200 or 300 Series meter able to edit Specific method if Password protected
Issue 
 Orion STARA200 /300 series meter  in  password-protected method can change the measurement mode?  (Example Conductivity to Salinity mode)
Environment
  • ORION STARA200/300 Series meter 
Resolution
  •    In the meter if the specific method is Password protected user unable to edit  without Password
  • At the same time meter allow  the user able to change the measurement mode without any issues (Example pH to mV or Conductivity to Salinity)
  • Analysis 
     Information has been verified by technical support team  with meter 
    Cause 
    Lacked  above details in the Meter instruction manual  
  • Was this article helpful?