S-STARA121-E 1213 - Rev B - Orion Star A121 pH Meter Product Specifications
The Thermo Scientific Orion Star A121 pH Handheld Meter offers precise pH and mV measurements with user-friendly features. It includes a large LCD for clear readings, AUTO-READ™ stability indicator, up to 3-point pH calibration, and automatic buffer recognition. The device supports both AC and battery power, has a durable IP54-rated housing, and comes with various kits and accessories for enhanced functionality. Ideal for laboratory use, it provides reliable data logging and easy calibration.
Description:
Product specification sheet for STARA121.
Environment:
- STARA121
Attachment(s):
| File | Last Modified |
|---|---|
| S-STARA121-E 1213 - Rev B - Orion Star A121 pH Meter Product Specifications.pdf | March 05, 2026 |