IS-VSSOFTWARE-E - Rev A - Orion Versa Star Meter - Software Update Procedure
The article provides instructions for updating Thermo Scientific Orion Versa Star meter software. It requires administrative access, Windows XP, Vista, or 7, and .NET framework 4.0 or higher. Steps include downloading and extracting the software, connecting the meter to a computer via USB, and following specific procedures to complete the update. It emphasizes backing up data and offers contact information for technical support and additional resources.
Description:
Software update procedure
Environment:
Thermo Scientific™ Orion™ Versa Star™ meters
Attachment(s):
| File | Last Modified |
|---|---|
| IS-VSSOFTWARE-E - Rev A - Orion Versa Star Meter - Software Update Procedure.pdf | January 07, 2022 |