36100124 - Rev B - Precision General Purpose Water Baths - Installation and Service Manual
Description:
Installation and Service Manual
Environment:
General Purpose Water Baths models:
- 180
- 181
- 182
- 183
- 184
- 185
- 186
- 188
Catalog Number | Model Number | Electrical Characteristics | Capacity | |||
Volts | Watts | Amps | Liters | Gallons | ||
51221070 | 180 | 120 | 225 | 1.9 | 1.5 | 0.4 |
51221071 | 180 | 230 | 225 | 1.0 | 1.5 | 0.4 |
51221064 | 181 | 120 | 225 | 1.9 | 2.5 | 0.7 |
51221211 | 181 | 230 | 225 | 1.0 | 2.5 | 0.7 |
51221073 | 182 | 120 | 300 | 2.5 | 5.5 | 1.5 |
51221068 | 182 | 230 | 300 | 1.3 | 5.5 | 1.5 |
51221060 | 183 | 120 | 400 | 3.3 | 12.0 | 3.2 |
51221062 | 183 | 230 | 400 | 1.7 | 12.0 | 3.2 |
51221075 | 184 | 120 | 600 | 5.0 | 19.5 | 5.2 |
51221069 | 184 | 230 | 600 | 2.6 | 19.5 | 5.2 |
51221065 | 185 | 120 | 600 | 5.0 | 18.0 | 4.9 |
51221066 | 185 | 230 | 600 | 2.6 | 18.0 | 4.9 |
51221072 | 186 | 120 | 1200 | 10.0 | 43.0 | 11.4 |
51221074 | 186 | 230 | 1200 | 5.2 | 43.0 | 11.4 |
51221061 | 188 | 120 | 800 | 6.7 | 12.0 | 3.2 |
51221063 | Dual | 230 | 800 | 3.5 | Both Pans | Both Pans |
Attachment(s):
File | Last Modified |
---|---|
36100124 - Rev B - Precision General Purpose Water Baths - Installation and Service Manual.pdf | December 01, 2021 |