36100126 - Rev B - Precision Dubnoff and Shallow Form Shaking Water Baths - Installation and Service Manual
The Precision Dubnoff and Shallow Form Shaking Water Baths Installation and Service Manual (document 36100126 - Rev B) provides comprehensive guidance for the installation, operation, and maintenance of these water baths. This manual ensures users can effectively install, operate, and maintain their Precision Dubnoff and Shallow Form Shaking Water Baths.
Description:
Installation/Service Manual
Environment:
Precision Dubnoff and Shallow Form Shaking Water Baths
| Cat.No. | Description | Volts | Hertz | Watts | Amps |
| 51221076 | Dubnoff Metabolic Shaking Incubator Bath | 120 | 50/60 | 1000 | 8.3 |
| 51221077 | Dubnoff Metabolic Shaking Incubator Bath | 230 | 50/60 | 1000 | 4.3 |
| 51221079 | Shallow Form Shaking Bath | 120 | 50/60 | 1000 | 8.3 |
| 51221078 | Shallow Form Shaking Bath | 230 | 50/60 | 1000 | 4.3 |
Attachment(s):
| File | Last Modified |
|---|---|
| 36100126 - Rev B - Precision Dubnoff and Shallow Form Shaking Water Baths - Installation and Service Manual.pdf | December 01, 2021 |