ANWPGENUVUF 0614 - Thermo Scientific Barnstead GenPure UV-UF-TOC Water Purification System - Trace Metal Analysis of Water Produced
This article covers trace metal analysis of water from the Thermo Scientific Barnstead GenPure UV/UF-TOC system. Testing 40 elements with ICP-MS and IC found 37 below detection levels. It highlights effective purification for ultrapure water, recommending UV-only systems for highly sensitive tasks and UV plus ultrafilter systems for biological experiments. The study confirms the system's ability to meet TOC specifications, stressing careful configuration to prevent contamination.
Description:
Trace Metal analysis of water produced
Environment:
Thermo Scientific Barnstead GenPure UV/UF-TOC Water Purification System
Attachment(s):
| File | Last Modified |
|---|---|
| ANWPGENUVUF 0614 - Thermo Scientific Barnstead GenPure UV_UF-TOC Water Purification System - Trace Metal Analysis of Water Produced.pdf | January 07, 2022 |