ANWPGPNUEN 1214 - Thermo Scientific Barnstead GenPure UV UF Water Purification System - On-demand Nuclease- and Endotoxin-Free Lab Water
The article evaluates the Thermo Scientific Barnstead GenPure Pro UV/UF water purification system in producing nuclease- and endotoxin-free ultrapure water. It explains how the system uses ultrafiltration and UV light to eliminate RNase, DNase, and endotoxins. The study confirmed the system's effectiveness in reducing impurities below detectable limits, ensuring high-quality water for lab applications and enhancing efficiency and accuracy.
Description:
On-demand Nuclease- and Endotoxin-Free lab water
Environment:
Thermo Scientific Barnstead GenPure UV/UF Water Purification System
Attachment(s):
| File | Last Modified |
|---|---|
| ANWPGPNUEN 1214 - Thermo Scientific Barnstead GenPure UV_UF Water Purification System - On-demand Nuclease- and Endotoxin-Free Lab Water.pdf | January 07, 2022 |