Application Note - Trace Analysis of Volatile and Semi-volatile Organic Compounds in Water Produced from a Thermo Scientific Barnstead Genpure Pro UV Water Purification System
The document details an analysis of VOCs and S-VOCs in ultrapure water produced by the Thermo Scientific Barnstead GenPure Pro UV water purification system. The water met stringent ASTM Type 1 specifications and was tested using GCMS. Results showed VOC and S-VOC levels below detection limits, indicating the system's efficacy in producing high-purity water suitable for sensitive organic analyses. The system uses UV light and high-quality DI cartridges to achieve these results.
Description:
In this Article, the topic of trace organic levels are discussed with regard to ultrapure water. Covers techniques and best lab practices to help achieve consistent trace level results.
Environment:
50131956 GenPure Pro Basic system
50131954 GenPure Pro with UF option
50131952 GenPure Pro with UV option
50131950 GenPure Pro with UV and UF options
50131948 GenPure Pro with TOC and UV options
50131922 GenPure Pro with TOC, UV and UF options
Attachment(s):
| File | Last Modified |
|---|---|
| GenPure Pro Trace Organics App Note 0415 3.pdf | November 26, 2024 |