Thermo Fisher Scientific
Service Instructions - Aquanex Touch Smart Dispenser Operation Video
The video demonstrates the usage of the hand remote dispenser, Touch Smart Dispenser, featured on the Thermo Scientific Aquanex Ultrapure Water Purification System. It showcases the quick-view display, which provides information on tank level, dispenser ID, dispense volume setting, system health status, Type 1 water temperature, dispensing water purity, and dispense/flow speed selector buttons. This guide assists users in efficiently operating the dispenser and understanding its functionalities
Topic
Short video (no audio) instructions for operating the Aquanex™ Touch Smart Dispenser
Environment
  • 7101010  Aquanex 10L System with Dispenser
  • 7101011  Aquanex 10L System with Dispenser and Pretreatment
  • 7201030  Aquanex 30L System with Dispenser and 30L tank
  • 7201031  Aquanex 30L System with Dispenser, 30L tank and Pretreatment
Video
  • Was this article helpful?