50158356 - Thermo Scientific Barnstead Pretreatment System - Quick Reference Guide 09-18
50157886 Barnstead Pretreatment System eliminates impurities in feed water that could damage reverse osmosis membranes. This article provides guidelines for cartridge replacement and pressure gauge monitoring. It outlines three key components: the 1-micron prefilter, hardness stabilizer, and carbon cartridge, with recommended replacement intervals.
Description:
Quick reference guide for the three-filter Barnstead pretreatment system, used with RO-based systems, catalog number 50157886
Environment:
Article Number 50157886 Thermo Scientific Barnstead Pretreatment System
Attachment(s):
| File | Last Modified |
|---|---|
| 50158356 0918 - Thermo Scientific Barnstead Pretreatment System - Quick Reference Guide.pdf | January 05, 2022 |