60275 - D0820 ORGANICfree Cartridge - Installation Manual
This article provides instructions for cleaning, sanitizing, and installing the ORGANICfree cartridge in both recirculating and non-recirculating water purification systems. It emphasizes the importance of proper sanitization before installation, outlines the cartridge configuration for NANOpure II systems, and includes procedures for ensuring optimal water purity. It also recommends overnight soaking for non-recirculating systems and a minimum flush of ten liters after periods of nonuse
Description:
Installation manual
Environment:
NANOpure II cartridge configuration:
| 4-Module | 3-Module | ||
| 1. Pretreatment | D0836 | 1.Pretreatment | D0836 |
| 2. High Capacity | D0803 | 2. Ultrapure | D0809 |
| 3. Ultrapure | D0809 | 3. ORGANICfree | D0820 |
| 4. ORGANICfree | D0820 | ||
Attachment(s):
| File | Last Modified |
|---|---|
| 60275 - D0820 ORGANICfree Cartridge - Installation Manual.pdf | January 04, 2022 |