7991048 - Rev 1 - Level Monitor - Dual Acting - Series 2112 - Unit Installation
The Installation Guide for the Dual Acting Level Monitor covers installation, programming, and technical details. It monitors water levels in storage tanks, functioning as a direct-acting unit to prevent over-filling or as a reverse-acting unit to protect equipment from low water conditions. It includes a controller/indicator with a pressure plate mounted electrical switch and a U-tube assembly. The Level Monitor operates as a pressure-controlled electrical switch.
Description:
Unit installation
Environment:
Level Monitor - Dual Acting, series 2112
Attachment(s):
| File | Last Modified |
|---|---|
| 7991048 - Rev 1 - Level Monitor - Dual Acting, Series 2112 - Unit Installation.pdf | January 05, 2022 |