927216 - Rev 1 - MegaPure 11A Water Still Series 677 - Operating Manual
The Thermo Scientific MEGA-PURE 11A Water Still is a compact, all-glass unit producing 11 liters per hour of distilled water, suitable for various applications requiring non-pyrogenic water with high resistivity. It operates at 240V or 208V. The manual covers installation, operation, maintenance, troubleshooting, and replacement parts, including electrical requirements, water supply, unpacking, assembly, and servicing protocols
Description:
Operating manual and parts list
Environment:
MegaPure 11A Water Still, series 677:
| Model number | Voltage |
| A440117 (6745) | 208V |
| A440118 (6746) | 240V |
Attachment(s):
| File | Last Modified |
|---|---|
| 927216 - Rev 1 - MegaPure 11A Water Still Series 677 - Operating Manual.pdf | January 18, 2022 |