927500 - 7006757 - Rev 2 - Automatic Collection System Series 683 - Operating Manual
The manual for the Thermo Automatic Collection System (ACS) Series 683 outlines its use with MEGA-PURE® Water Stills. It covers safety, installation, assembly, maintenance, parts list, and ordering. The ACS stores up to 12 gallons (45 liters) of distilled water and signals the Water Still to replenish when levels drop to 35 liters, ensuring an automatic supply. It features a control knob and delivery spout for easy access.
Description:
Operating manual and parts list
Environment:
Automatic Collection System Series 683, model number B440704 (6757)
Attachment(s):
| File | Last Modified |
|---|---|
| 927500 (7006757) - Rev 2 - Automatic Collection System Series 683 - Operating Manual.pdf | January 04, 2022 |