Aquanex Water Purification System - Troubleshooting Blank Display and Clicking Noise
This article addresses the issue of clicking sounds and no display upon startup of the Aquanex water purification unit. The root cause is identified as the incorrect installation of the power cable. To resolve this, reinstall the plug so the flat side faces the USB connection. Follow the operation manual's guidelines to ensure proper positioning and avoid damage to the connector.
Description:
If the Aquanex water purification system makes clicking sounds and there is no display when plugged in, please check the attachement of the power cable.
The flat side of the plug should face inward, with the flat side toward the USB port. In some cases it has been installed backwards which creates the symptoms described.
Environment:
7101010 Aquanex 10L System with Dispenser
7101011 Aquanex 10L System with Dispenser and Pretreatment
7201030 Aquanex 30L System with Dispenser and 30L tank
7201031 Aquanex 30L System with Dispenser, 30L tank and Pretreatment
Attachment(s):
| File | Last Modified |
|---|---|
| Aquanex Water Purification System clicking noise upon start up no display V1.pdf | February 25, 2025 |