E-Pure Water Purification Systems - Troubleshooting Guide for Leaking Cartridge Holder - Nov 2024
This article provides best practices for installing canisters in the E-pure system to prevent leaks. Key steps include correctly positioning O-rings, securing canisters, and ensuring proper water supply. It also offers troubleshooting tips for persistent leaks, emphasizing the importance of using Thermo Scientific consumables and verifying proper installation.
Description:
Troubleshooting information for leaking E-Pure system. Use for troubleshooting canister leaks in D4641 and D4631 water purification systems.
Includes information to correctly install o-ring, canister and handle to avoid leaks.
Environment:
D4641
D4631
Attachment(s):
| File | Last Modified |
|---|---|
| E-Pure Water Purification Systems - Troubleshooting Guide for Leaking E-Pure System V2.pdf | November 05, 2024 |