Thermo Fisher Scientific
Harvey DI+ Cartridge System Model AY1273X4 - Operation Manual and Parts List - LT1273X13
The attached operator manual covers the Thermo Scientific Harvey DI+ Cartridge System. It includes guidance on operation, maintenance, system components, and troubleshooting. It serves as a training resource for new operators and maintenance personnel, ensuring safe and effective use. The manual aims to maximize system efficiency and longevity through detailed instructions and regular maintenance schedules, enhancing the performance of the water purification system.
Description: 
Instruction manual for users of Series 1273 Thermo Scientific Harvey DI+ Cartridge System, catalog number AY1273X4. The unit includes a remote dispenser, Barnstead Pura-Lite indicator, and a D0803 high-capacity cartridge. The system removes calcium, magnesium, and other ions, with a flow rate of up to 4 L/min. It features a pressure regulator, wall-mounting capability, and easy maintenance with a quarter-turn release canister.
Environment:
Harvey™ DI+ Cartridge System with dispenser, pura-lite, cartridge 
catalog numberAY1273X4