Installation and Operating Instructions for Barnstead Hose Nipple Cartridges 60031 - 3/10/08
Description:
This is the Installation and Operating Instruction Sheet for Barnstead Hose Nipple Cartridges. These cartridges are for use with the simple D8900 wall-mount cartridge holder. Designed for non-pressure applications, these cartridges include 3/8 in. barb hose connections on each end. They provide the ability to see when resin is exhausted in specific cartridges by a color change indicator.
Environment:
D8901 High Capacity
D8902 Ultrapure (mixed resin)
D8903 Oxygen Removal
D8904 Organic Removal
D8905 Cation Removal
D8921 Scale Eliminator
D8922 Combination
D8950 Economy High Capacity
D8908 LL Cation Removal
D8910 Acid Fume Absorption
D8911 Ultrapure no dye
D8911-X001 Special Ultrapure no dye
D8951 Economy
D8822 Economy mixed bed/oxygen
D50217 Combination Oxygen Removal & mixed bed
Attachment(s):
| File | Last Modified |
|---|---|
| USER MANUAL - BARNSTEAD - 60031 - DEIONIZATION - HOSE NIPPLE CARTRIDGE INSTALLATION.PDF | February 19, 2025 |