LT1332X1 - Rev 3 - EASYpure RoDi Ultrapure Water Purification System Series 1332 - Operating Manual
This manual outlines the operation and parts list for the Barnstead EASYpure RODI Ultrapure Water Purification System, Series 1332. It details how the system uses reverse osmosis and deionization processes to produce reagent grade water exceeding ASTM and NCCLS/CAP Type I standards. The system features automatic RO operation, continuous DI water resistivity monitoring, and a self-contained 6.5-liter reservoir. Essential components must be ordered separately.
Description:
Operating manual and parts list
Environment:
EASYpure® RoDi Ultrapure Water Purification System Series 1332:
| Model number | Voltage |
| D13321 (7128) | 100-240V |
Attachment(s):
| File | Last Modified |
|---|---|
| LT1332X1 - Rev 3 - EASYpure RoDi Ultrapure Water Purification System Series 1332 - Operating Manual.pdf | January 04, 2022 |