LT1923X1 - 7006749 - Rev 1 - MegaPure 3A Water Still Series 1923 - Operating Manual
The Thermo Scientific MEGA-PURE 3A Water Still is a compact, all-glass unit producing 3 liters per hour of distilled water, suitable for various applications requiring non-pyrogenic water with high resistivity. It operates at 240V or 208V. The manual covers installation, operation, maintenance, troubleshooting, and replacement parts, including electrical requirements, water supply, unpacking, assembly, and servicing protocols
Description:
Operating manual and parts list
Environment:
MegaPure 3A Water Still, series 1923:
| Model number | Voltage |
| A440696 (6751) | 208V |
| A440367 (6749) | 220-240V |
MP3A
Attachment(s):
| File | Last Modified |
|---|---|
| LT1923X1 (7006749) - Rev 1 - MegaPure 3A Water Still Series 1923 - Operating Manual.pdf | January 18, 2022 |