Thermo Fisher Scientific
LT2112X1 (7006769) - Rev 1 - Classic Electrically Heated Stills Series 2112 - Operating Manual
Description: 
Operating manual and parts list 
Environment:
Classic Electrically Heated Stills Series 2112, model numbers:
Model Number Voltage Size
A1011-A (6768) 120V, 1PH 1 gallon
A1011-B-61 (6769) 240V, 1PH 1 gallon
A1013-C (6771) 120/208V, 3PH 2 gallon
A1013-B-61 (6786) 240V, 1PH 2 gallon
A1015-B-61 (6772) 240V, 1 PH 5 gallon
A1015-C (6773) 120/208V, 3PH 5 gallon
A1016-D (6774) 240V, 3PH 10 gallon