Operator Manual - D8952 Thermo Scientific Barnstead Remote Dispenser - LT895X2 Rev 0 - 8/27/10
This article provides the operator instruction manual for the Barnstead Remote Dispenser (part number D8952). This optional accessory delivers Type I reagent grade water up to 8 feet (2.4 meters) from an existing Epure water system and is compatible with obsolete NANOpure Quad systems. The remote dispenser includes all necessary parts and hardware for installation, a final filter, and a mounting bracket.
Description:
Instructions for installation and operation of D8952 remote dispenser as used on E-Pure water purification systems.
Also compatible older Nanopure Quad water systems, which are no longer supported.
Environment
E-Pure model D4641
E-Pure model D4631
Attachment(s):
| File | Last Modified |
|---|---|
| USER MANUAL - REMOTE DISPENSER - TYPE D8952 - SERIES 985 - LT895X2 - 327142 REV. 0.pdf | April 29, 2025 |