Operator Manual - Thermo Scientific VENTGUARD storage tank accessory
This operation manual and parts list for Ventgard® and Water Seal describes their installation and maintenance for preventing contamination in distilled water storage tanks. The Ventgard uses breather valves and a filter element to purify air entering the tank, while the Water Seal prevents air from entering the tank overflow. Maintenance includes replacing the Ventgard filter element every 60 days or after 1,000 gallons.
Description:
User Manual for the Ventguard accessory, H3120 used on the storage tank of classic stills and Biopure stills. Used to prevent the distilled water in the storage
tanks from being contaminated with airborne impurities, Incudes parts list with gaskets, filter element and more.
Environment:
A1065-B (6759) 120/240, 1 phase
A1065-C (6760) 120/208V, 3 phase
A1065-D (6761) 120/240V, 3 phase
A1011-A (6768) 120V, 1PH, 1 gallon
A1011-B-61 (6769) 240V, 1PH, 1 gallon
A1013-C (6771) 120/208V, 3PH, 2 gallon
A1013-B-61 (6786) 240V, 1PH, 2 gallon
A1015-B-61 (6772) 240V, 1 PH ,5 gallon
A1015-C (6773) 120/208V, 3PH, 5 gallon
A1016-D (6774) 240V, 3PH, 10 gallon
Attachment(s):
| File | Last Modified |
|---|---|
| 25001-DB discontinuation - communication.pdf | November 04, 2025 |
| USER MANUAL - BARNSTEAD - VENTGARD AND WATER SEAL - 60222.pdf | May 30, 2024 |