Service Instruction - E-Pure remote dispenser connection and water flow information
Epure Flow Path with Remote Dispenser Installation Issue - When installing the remote dispenser, ensure the recirculation line is moved to the back bottom connection and the plug to the upper back connection. Incorrect installation can cause water leaks or recirculation failure. Applicable for E-Pure models D4641 and D4631 with Remote Dispenser D8952. Use the provided pictures and instructions to confirm proper installation.
Description: Environment
This instructional document covers installation questions regarding E-Pure systems. It is helpful to customers and service technicians who are installing or troubleshooting D4641 & D4631 water purification systems. It has photos and information about the connections of the recirculation tubing and the optional remote dispenser. It also includes supplemental instructions for installing the optional remote dispenser, part number D8952.
Environment
E-Pure model D4641
E-Pure Model D4631
Remote Dispenser D8952
Attachment(s):
| File | Last Modified |
|---|---|
| Epure Flow Path with Remote Dispenser Rev 1.pdf | September 30, 2025 |