Smart2Pure Water Purification Systems - Storage Reservoir Will Not Fill
The Troubleshooting Guide for Smart2Pure systems addresses storage reservoir fill issues. Causes include feed water issues, blocked membranes/cartridges, incorrect concentrate settings, float switch problems, solenoid/check valve failures, low pressure switches, and RO boost pump issues. The guide offers corrective actions for each cause to restore function and ensure proper reservoir filling.
Description:
Troubleshooting guide. Occasionally the storage reservoir may not fill in a Smart2Pure water purification system.
Environment:
Smart2Pure water purification systems
Attachment(s):
| File | Last Modified |
|---|---|
| Smart2Pure Water Purification Systems - Storage Reservoir Will Not Fill.pdf | January 05, 2022 |