Spec Sheet - Harvey DI+ Cartridge System Model AY1273X4
This article includes the brochure for Thermo Scientific Harvey DI+ Cartridge System provides quick pure water for lab use, humidification, and steam autoclaves. The DI Plus is a turn-key system including a remote dispenser, Barnstead Pura-Lite indicator, and a high-capacity, two-bed ion exchange cartridge to remove calcium, magnesium, and other ions. It supports various cartridges, delivering pure water at up to 4 L/min. Features include a pressure regulator and compact wall-mount design
Description:
Brochure for AY1273X4 Thermo Scientific Harvey DI+ Cartridge System. The system is a turn-key solution for producing pure water for various lab applications. It includes a remote dispenser, Barnstead Pura-Lite indicator, and a high-capacity ion exchange cartridge. The system removes calcium, magnesium, and other ions, with a flow rate of up to 4 L/min. It features a pressure regulator, wall-mounting capability, and easy maintenance with a quarter-turn release canister.
Environment:
Harvey™ DI+ Cartridge System with dispenser, pura-lite, cartridge
catalog numberAY1273X4
Attachment(s):
| File | Last Modified |
|---|---|
| HarveyDI-CartridgeSystem-SpecSheet-EN.pdf | May 02, 2025 |